1-Docosanethiol ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Docosyl Mercaptan

D3905-5G
TCD3905-5GEA 222.22 USD
TCD3905-5G
1-Docosanethiol ≥98.0% (by GC, titration analysis)
1-Docosanethiol

CAS Number: 7773-83-3
MDL Number: MFCD02262163
Molecular Formula: C22H46S
Molecular Weight: 342.67
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 48


Other Product Names / Synonyms: Docosyl Mercaptan

Formula: C₂₂H₄₆S
MW: 342.67 g/mol
Melting Pt: 48 °C
MDL Number: MFCD02262163
CAS Number: 7773-83-3
https://us.vwr.com/stibo/web/std.lang.all/58/52/28985852.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR