2,5-Dihydroxyterephthalic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D3899-5G
D3899-25G
TCD3899-25GEA
394.54
USD
TCD3899-25G
TCD3899-5G
2,5-Dihydroxyterephthalic acid ≥98.0% (by HPLC, titration analysis)
2,5-Dihydroxyterephthalic acid
CAS Number: 610-92-4
MDL Number: MFCD00132933
Molecular Formula: C8H6O6
Molecular Weight: 198.13
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 339
Formula:
C₈H₆O₆ MW: 198.13 g/mol Melting Pt: 339 °C |
MDL Number:
MFCD00132933 CAS Number: 610-92-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...