Di-tert-butyl-L-(+)-tartrate ≥97.0% (by GC)
Supplier: TCI America
Synonyms:
L-(+)-Tartaric Acid Di-tert-Butyl Ester
D3898-1G
TCD3898-1GEA
273.97
USD
TCD3898-1G
Di-tert-butyl-L-(+)-tartrate ≥97.0% (by GC)
Di-tert-butyl-L-(+)-tartrate
CAS Number: 117384-45-9
MDL Number: MFCD00192000
Molecular Formula: C12H22O6
Molecular Weight: 262.30
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 90
Specific rotation [a]20/D: 11 deg (C=1, Acetone)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: L-(+)-Tartaric Acid Di-tert-Butyl Ester
Formula:
C₁₂H₂₂O₆ MW: 262.30 g/mol Melting Pt: 90 °C |
MDL Number:
MFCD00192000 CAS Number: 117384-45-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...