2,9-Diphenyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D3849-1G
TCD3849-1GEA
432.37
USD
TCD3849-1G
2,9-Diphenyl-1,10-phenanthroline ≥98.0% (by HPLC, titration analysis)
2,9-Diphenyl-1,10-phenanthroline
CAS Number: 25677-69-4
Molecular Formula: C24H16N2
Molecular Weight: 332.41
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 188
Formula:
C₂₄H₁₆N₂ MW: 332.41 g/mol Melting Pt: 188 °C |
MDL Number:
MFCD00044208 CAS Number: 25677-69-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...