Di-tert-butylsilane ≥95.0% (by GC)

Supplier: TCI America

D3731-1G D3731-5G
TCD3731-5GEA 229.01 USD
TCD3731-5G TCD3731-1G
Di-tert-butylsilane ≥95.0% (by GC)
Di-tert-butylsilane

CAS Number: 30736-07-3
MDL Number: MFCD00270996
Molecular Formula: C8H20Si
Molecular Weight: 144.33
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Melting point (°C): -38
Flash Point (°C): -2
Specific Gravity (20/20): 0.73

https://us.vwr.com/stibo/web/std.lang.all/57/04/28985704.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR