Naphthalene-2,7-dicarbonitrile ≥94.0% (by GC)

Supplier: TCI America

D3705-1G
TCD3705-1GEA 177 USD
TCD3705-1G
Naphthalene-2,7-dicarbonitrile ≥94.0% (by GC)
Naphthalene-2,7-dicarbonitrile

CAS Number: 39718-11-1
Molecular Formula: C12H6N2
Molecular Weight: 178.19
Purity/Analysis Method: 94.0% (GC)
Form: Crystal
Melting point (°C): 267


Other Product Names / Synonyms: 2,7-Naphthalenedicarbonitrile

https://us.vwr.com/stibo/web/std.lang.all/56/82/28985682.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR