3,4-Difluorophenyl isocyanate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Isocyanic acid 3,4-Difluorophenyl ester
CAS Number: 42601-04-7
MDL Number: MFCD00065711
Molecular Formula: C7H3F2NO
Molecular Weight: 155.10
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 85
Flash Point (°C): 63
Specific Gravity (20/20): 1.34
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Isocyanic Acid 3,4-Difluorophenyl Ester
Formula:
C₇H₃F₂NO MW: 155.1 g/mol Boiling Pt: 85 °C Density: 1.34 (20 °C) Flash Pt: 63 °C |
MDL Number:
MFCD00065711 CAS Number: 42601-04-7 UN: 2206 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...