1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 1,3-Di(adamantan-1-yl)-1H-imidazol-3-ium tetrafluoroborate

D3621-1G
TCD3621-1GEA 162.8 USD
TCD3621-1G
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate ≥96.0% (by total nitrogen basis)
1,3-Bis(1-adamantyl)imidazolium tetrafluoroborate

CAS Number: 286014-42-4
MDL Number: MFCD04973311
Molecular Formula: C23H33BF4N2
Molecular Weight: 424.33
Purity/Analysis Method: 96.0% (N)
Form: Crystal
Melting point (°C): 282


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, Ligands
Other Product Names / Synonyms: IAd.HBF4

Formula: C₂₃H₃₃BF₄N₂
MW: 424.33 g/mol
Melting Pt: 282 °C
MDL Number: MFCD04973311
CAS Number: 286014-42-4
https://us.vwr.com/stibo/web/std.lang.all/56/10/28985610.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR