α-Bromo-2,5-dichlorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(Bromomethyl)-1,4-dichlorobenzene2,5-Dichlorobenzyl bromide

D3548-25G D3548-5G
TCD3548-5GEA 76 USD
TCD3548-5G TCD3548-25G
α-Bromo-2,5-dichlorotoluene ≥98.0% (by GC)
α-Bromo-2,5-dichlorotoluene

CAS Number: 85482-13-9
MDL Number: MFCD00236034
Molecular Formula: C7H5BrCl2
Molecular Weight: 239.92
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 138
Melting point (°C): 42


Other Product Names / Synonyms: 1-(Bromomethyl)-2,5-dichlorobenzene, alpha-Bromo-2,5-dichlorotoluene

Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Boiling Pt: 138 °C
Melting Pt: 42 °C
MDL Number: MFCD00236034
CAS Number: 85482-13-9
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/06/20/28980620.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR