9-(2,5-Dimethylphenyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC)

D3428-5G D3428-1G
TCD3428-1GEA 243.78 USD
TCD3428-1G TCD3428-5G
9-(2,5-Dimethylphenyl)-10-methylacridinium perchlorate ≥98.0% (by HPLC)
9-(2,5-Dimethylphenyl)-10-methylacridinium perchlorate

Molecular Weight: 397.86
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal
Molecular Formula: C22H20ClNO4

https://us.vwr.com/stibo/web/std.lang.all/04/24/28980424.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR