2,6-Difluoro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Cyano-3,5-difluorophenol

D3424-5G
TCD3424-5GEA 234 USD
TCD3424-5G
2,6-Difluoro-4-hydroxybenzonitrile ≥98.0% (by GC, titration analysis)
2,6-Difluoro-4-hydroxybenzonitrile

CAS Number: 123843-57-2
MDL Number: MFCD03094499
Molecular Formula: C7H3F2NO
Molecular Weight: 155.10
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 127


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-3,5-difluorophenol

Formula: C₇H₃F₂NO
MW: 155.1 g/mol
Melting Pt: 127 °C
MDL Number: MFCD03094499
CAS Number: 123843-57-2
https://us.vwr.com/stibo/web/std.lang.all/04/17/28980417.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR