Dimethyl trisulfide ≥98.0% (by GC)

Supplier: TCI America

D3418-25G D3418-5G
TCD3418-5GEA 37.54 USD
TCD3418-5G TCD3418-25G
Dimethyl trisulfide ≥98.0% (by GC)
Dimethyl trisulphide

CAS Number: 3658-80-8
MDL Number: MFCD00039808
Molecular Formula: C2H6S3
Molecular Weight: 126.25
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 170
Melting point (°C): -68
Flash Point (°C): 56
Specific Gravity (20/20): 1.21

Formula: C₂H₆S₃
MW: 126.27 g/mol
Boiling Pt: 170 °C
Melting Pt: –68 °C
Density: 1.21 (20 °C)
Flash Pt: 56 °C
MDL Number: MFCD00039808
CAS Number: 3658-80-8
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/04/10/28980410.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR