3,4-Dichlorobenzonitrile ≥98.0% (by GC)

Supplier: TCI America

D3415-25G D3415-5G
TCD3415-5GEA 22.33 USD
TCD3415-5G TCD3415-25G
3,4-Dichlorobenzonitrile ≥98.0% (by GC)
3,4-Dichlorobenzonitrile

CAS Number: 6574-99-8
MDL Number: MFCD00016379
Molecular Formula: C7H3Cl2N
Molecular Weight: 172.01
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 73

Formula: C₇H₃Cl₂N
MW: 172.01 g/mol
Melting Pt: 73 °C
MDL Number: MFCD00016379
CAS Number: 6574-99-8
UN: 3439
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/04/07/28980407.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR