2,3-Difluoroanisole ≥97.0% (by GC)

Synonyms: 1,2-Difluoro-3-methoxybenzene

D3359-5G D3359-1G
TCD3359-1GEA 14.57 USD
TCD3359-1G TCD3359-5G
2,3-Difluoroanisole ≥97.0% (by GC)
2,3-Difluoroanisole

CAS Number: 134364-69-5
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.24

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Density: 1.24 (20 °C)
MDL Number: MFCD01631562
CAS Number: 134364-69-5
https://us.vwr.com/stibo/web/std.lang.all/03/47/28980347.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR