ω-Bromo-2,4-dichloroacetophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromo-1-(2,4-dichlorophenyl)ethan-1-one2,4-Dichlorophenacyl bromideα-Bromo-2,4-dichloroacetophenone2-Bromo-2',4'-dichloroacetophenone2-Bromo-1-(2,4-dichlorophenyl)ethanone

D3351-5G
TCD3351-5GEA 105.56 USD
TCD3351-5G
ω-Bromo-2,4-dichloroacetophenone ≥98.0% (by GC)
ω-Bromo-2,4-dichloroacetophenone

CAS Number: 2631-72-3
MDL Number: MFCD00053005
Molecular Formula: C8H5BrCl2O
Molecular Weight: 267.93
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 24


Other Product Names / Synonyms: 2-Bromo-2',4'-dichloroacetophenone

Formula: C₈H₅BrCl₂O
MW: 267.94 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00053005
CAS Number: 2631-72-3
https://us.vwr.com/stibo/web/std.lang.all/03/37/28980337.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR