Dodecyl isocyanate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1-Dodecyl isocyanate

D3348-5G
TCD3348-5GEA 77.35 USD
TCD3348-5G
Dodecyl isocyanate ≥97.0% (by GC)
Dodecyl isocyanate

CAS Number: 4202-38-4
MDL Number: MFCD00041744
Molecular Formula: C13H25NO
Molecular Weight: 211.35
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 161
Flash Point (°C): 113
Specific Gravity (20/20): 0.87
Storage Temperature: 0-10°C


Other Product Names / Synonyms: Isocyanic Acid Dodecyl Ester

Formula: C₁₃H₂₅NO
MW: 211.35 g/mol
Boiling Pt: 161 °C
Density: 0.87 (20 °C)
Flash Pt: 113 °C
MDL Number: MFCD00041744
CAS Number: 4202-38-4
UN: 2206
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/03/34/28980334.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR