Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride ≥95.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Bis(pentamethylcyclopentadienyl)zirconium dichloride

D3307-1G
TCD3307-1GEA 117 USD
TCD3307-1G
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride ≥95.0% (by titrimetric analysis)
Bis(pentamethylcyclopentadienyl)zirconium(IV) dichloride

CAS Number: 54039-38-2
Molecular Formula: C20H30Cl2Zr
Molecular Weight: 432.58
Purity/Analysis Method: 95.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Polymerization Catalysts
Other Product Names / Synonyms: Bis(pentamethylcyclopentadienyl)zirconium(IV) Dichloride

Formula: C₂₀H₃₀Cl₂Zr
MW: 432.582  g/mol
MDL Number: MFCD00058849
CAS Number: 54039-38-2
UN: 3096
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/02/80/28980280.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR