1,4-Dibutoxybenzene ≥98.0% (by GC)

Supplier: TCI America

D3264-5G
TCD3264-5GEA 191 USD
TCD3264-5G
1,4-Dibutoxybenzene ≥98.0% (by GC)
1,4-Dibutoxybenzene

CAS Number: 104-36-9
Molecular Formula: C14H22O2
Molecular Weight: 222.33
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 158
Melting point (°C): 45

Formula: C₁₄H₂₂O₂
Boiling Pt: 158 °C
Melting Pt: 45 °C
CAS Number: 104-36-9
https://us.vwr.com/stibo/web/std.lang.all/02/49/28980249.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR