Dichloro(dimethylamino)phosphine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Dimethylphosphoramidous dichloride(Dimethylamino)phosphonous Dichloride

D3245-5G
TCD3245-5GEA 160.37 USD
TCD3245-5G
Dichloro(dimethylamino)phosphine ≥98.0% (by GC)
Dichloro(dimethylamino)phosphine

CAS Number: 683-85-2
Molecular Formula: C2H6Cl2NP
Molecular Weight: 145.95
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): -23
Specific Gravity (20/20): 1.26
Storage Temperature: 0-10°C


Other Product Names / Synonyms: (Dimethylamino)phosphonous Dichloride

Formula: C₂H₆Cl₂NP
MW: 145.95 g/mol
Boiling Pt: 150 °C
Density: 1.26 (20 °C)
Flash Pt: –23 °C
MDL Number: MFCD00013613
CAS Number: 683-85-2
https://us.vwr.com/stibo/web/std.lang.all/02/33/28980233.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR