Dicobalt octacarbonyl stabilized
Supplier: TCI America
Synonyms:
Cobalt Carbonyl
D3213-5G
D3213-25G
TCD3213-5GEA
181.59
USD
TCD3213-5G
TCD3213-25G
Dicobalt octacarbonyl stabilized
Dicobalt octacarbonyl
(stabilized with 1-5% Hexane)
CAS Number: 10210-68-1
MDL Number: MFCD00016024
Molecular Formula: C8Co2O8
Molecular Weight: 341.95
Form: Crystal
Color: Yellow Red
Melting point (°C): 51
Flash Point (°C): -13
Storage Temperature: 0-10°C
Other Product Names / Synonyms: Cobalt Carbonyl, Octacarbonyldicobalt
Formula:
C₈Co₂O₈ MW: 341.95 g/mol Melting Pt: 51 °C Flash Pt: –13 °C |
MDL Number:
MFCD00016024 CAS Number: 10210-68-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...