Diamyl sulfate ≥96.0% (by GC)
Supplier: TCI America
Synonyms:
Dipentyl Sulphate
, Dipentyl Sulfate
CAS Number: 5867-98-1
MDL Number: MFCD06797095
Molecular Formula: C10H22O4S
Molecular Weight: 238.34
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 117
Melting point (°C): 14
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification)
Other Product Names / Synonyms: Dipentyl Sulfate
Formula:
C₁₀H₂₂O₄S MW: 238.34 g/mol Boiling Pt: 117 °C Melting Pt: 14 °C Density: 1.03 (20 °C) |
MDL Number:
MFCD06797095 CAS Number: 5867-98-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...