Diisopropyl sulfate ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Diisopropyl SulfateDiisopropyl sulphate

D2954-25G D2954-5G
TCD2954-25GEA 589.55 USD
TCD2954-25G TCD2954-5G
Diisopropyl sulfate ≥97.0% (by GC)
Diisopropyl sulfate

CAS Number: 2973-10-6
MDL Number: MFCD01682898
Molecular Formula: C6H14O4S
Molecular Weight: 182.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 94
Melting point (°C): -19
Specific Gravity (20/20): 1.10
Storage Temperature: 0°C

Formula: C₆H₁₄O₄S
MW: 182.23 g/mol
Boiling Pt: 94 °C
Melting Pt: –19 °C
Density: 1.10 (20 °C)
MDL Number: MFCD01682898
CAS Number: 2973-10-6
https://us.vwr.com/stibo/web/std.lang.all/00/97/28980097.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR