Diphosphoryl tetrachloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Pyrophosphoryl chloride
D2883-25G
TCD2883-25GEA
120.8
USD
TCD2883-25G
Diphosphoryl tetrachloride ≥98.0% (by titrimetric analysis)
Diphosphoryl tetrachloride
CAS Number: 13498-14-1
MDL Number: MFCD00061542
Molecular Formula: Cl4O3P2
Molecular Weight: 251.74
Purity/Analysis Method:
Form: Clear Liquid
Specific Gravity (20/20): 1.82
Product Categories: Synthetic Organic Chemistry, Phosphorylation, Nucleic Acid Synthesis
Other Product Names / Synonyms: Pyrophosphoryl Chloride
Formula:
Cl₄O₃P₂ MW: 251.76 g/mol Density: 1.82 (20 °C) |
MDL Number:
MFCD00061542 CAS Number: 13498-14-1 UN: 3265 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...