1,5-Dimethoxypentane ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1,5-Pentanediol dimethyl etherPentamethylene glycol dimethyl ether

D2857-5G
TCD2857-5GEA 66.99 USD
TCD2857-5G
1,5-Dimethoxypentane ≥97.0% (by GC)
1,5-Dimethoxypentane

CAS Number: 111-89-7
MDL Number: MFCD03613608
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid


Other Product Names / Synonyms: Pentamethylene Glycol Dimethyl Ether, 1,5-Pentanediol Dimethyl Ether

https://us.vwr.com/stibo/web/std.lang.all/00/12/28980012.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR