3,4-Difluorobenzoyl chloride ≥95.0% (by GC)

Synonyms: 3,4-difluorobenzoylchloride

D2847-5G D2847-25G
TCD2847-25GEA 188.7 USD
TCD2847-25G TCD2847-5G
3,4-Difluorobenzoyl chloride ≥95.0% (by GC)
3,4-Difluorobenzoyl chloride

CAS Number: 76903-88-3
MDL Number: MFCD00009930
Molecular Formula: C7H3ClF2O
Molecular Weight: 176.55
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 44
Flash Point (°C): 78
Specific Gravity (20/20): 1.51

Formula: C₇H₃ClF₂O
MW: 176.55 g/mol
Boiling Pt: 44 °C
Density: 1.51 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00009930
CAS Number: 76903-88-3
UN: 3265
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/00/03/28980003.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR