Diphenylacetyl chloride ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2,2-Diphenylacetyl chloride
D2763-25G
D2763-5G
TCD2763-25GEA
169.51
USD
TCD2763-25G
TCD2763-5G
Diphenylacetyl chloride ≥98.0% (by GC, titration analysis)
Diphenylacetyl chloride
CAS Number: 1871-76-7
MDL Number: MFCD00013655
Molecular Formula: C14H11ClO
Molecular Weight: 230.69
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 53
Flash Point (°C): 110
Formula:
C₁₄H₁₁ClO MW: 230.69 g/mol Melting Pt: 53 °C Flash Pt: 110 °C |
MDL Number:
MFCD00013655 CAS Number: 1871-76-7 UN: 3261 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...