Naphthalene-1,4-dicarbonitrile ≥98.0% (by GC)

Supplier: TCI America

D2758-5G
TCD2758-5GEA 159.36 USD
TCD2758-5G
Naphthalene-1,4-dicarbonitrile ≥98.0% (by GC)
Naphthalene-1,4-dicarbonitrile

CAS Number: 3029-30-9
MDL Number: MFCD03093626
Molecular Formula: C12H6N2
Molecular Weight: 178.19
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 208


Other Product Names / Synonyms: 1,4-Naphthalenedicarbonitrile

Formula: C₁₂H₆N₂
Melting Pt: 208 °C
MDL Number: MFCD03093626
CAS Number: 3029-30-9
https://us.vwr.com/stibo/web/std.lang.all/98/96/28979896.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR