2-Chloromethyl-p-xylene ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
α-Chloro-1,2,5-trimethylbenzene
, 2,5-Dimethylbenzyl chloride
, 2-(Chloromethyl)-1,4-dimethylbenzene
, 1-(Chloromethyl)-2,5-dimethylbenzene
D2716-25G
TCD2716-25GEA
150
USD
TCD2716-25G
2-Chloromethyl-p-xylene ≥98.0% (by GC, titration analysis)
2-Chloromethyl-p-xylene
CAS Number: 824-45-3
MDL Number: MFCD00000902
Molecular Formula: C9H11Cl
Molecular Weight: 154.64
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 65
Specific Gravity (20/20): 1.06
Other Product Names / Synonyms: 1-Chloromethyl-2,5-dimethylbenzene
Formula:
C₉H₁₁Cl MW: 154.64 g/mol Boiling Pt: 121 °C Density: 1.06 (20 °C) Flash Pt: 65 °C |
MDL Number:
MFCD00000902 CAS Number: 824-45-3 UN: 3265 ADR: 8,II |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...