Dinonylamine ≥97.0% (by GC)

Supplier: TCI America

D2676-5G
TCD2676-5GEA 358.19 USD
TCD2676-5G
Dinonylamine ≥97.0% (by GC)
Dinonylamine

CAS Number: 2044-21-5
MDL Number: MFCD00027338
Molecular Formula: C18H39N
Molecular Weight: 269.52
Purity/Analysis Method: 97.0% (GC)
Form: Crystal
Freezing point (°C): 24

Formula: C₁₈H₃₉N
MDL Number: MFCD00027338
CAS Number: 2044-21-5
https://us.vwr.com/stibo/web/std.lang.all/96/89/28979689.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR