α-Bromo-2,5-difluorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2,5-Difluorobenzyl bromide

D2597-5G D2597-25G
TCD2597-25GEA 113.68 USD
TCD2597-25G TCD2597-5G
α-Bromo-2,5-difluorotoluene ≥98.0% (by GC)
α-Bromo-2,5-difluorotoluene

CAS Number: 85117-99-3
MDL Number: MFCD00009897
Molecular Formula: C7H5BrF2
Molecular Weight: 207.02
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 15
Specific Gravity (20/20): 1.64


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: alpha-Bromo-2,5-difluorotoluene

Formula: C₇H₅BrF₂
MW: 207.02 g/mol
Density: 1.64 (20 °C)
Flash Pt: 15 °C
MDL Number: MFCD00009897
CAS Number: 85117-99-3
UN: 3265
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/95/66/28979566.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR