Methyl diphenylphosphinite ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Methoxydiphenylphosphine
D2557-5G
TCD2557-5GEA
107
USD
TCD2557-5G
Methyl diphenylphosphinite ≥98.0% (by GC)
Methyl diphenylphosphinite
CAS Number: 4020-99-9
MDL Number: MFCD00048025
Molecular Formula: C13H13OP
Molecular Weight: 216.22
Purity/Analysis Method:
Form: Clear Liquid
Flash Point (°C): 113
Specific Gravity (20/20): 1.10
Product Categories: Synthetic Organic Chemistry, Ligands
Other Product Names / Synonyms: Diphenylphosphinous Acid Methyl Ester, Methyl Diphenylphosphinite
Formula:
C₁₃H₁₃OP MW: 216.22 g/mol Density: 1.10 (20 °C) Flash Pt: 113 °C |
MDL Number:
MFCD00048025 CAS Number: 4020-99-9 |
![]() |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...