(S)-N,N-Dimethyl-1-[(R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine ≥95.0% (by GC)

Supplier: TCI America

Synonyms: (S)-(R)-BPPFA

D2538-100MG
TCD2538-100MGEA 81.17 USD
TCD2538-100MG
(S)-N,N-Dimethyl-1-[(R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine ≥95.0% (by GC)
(S)-N,N-Dimethyl-1-[(R)-1',2-bis(diphenylphosphino)ferrocenyl]ethylamine

CAS Number: 55650-59-4
MDL Number: MFCD00075283
Molecular Formula: C38H37FeNP2
Molecular Weight: 625.51
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 142
Specific rotation [a]20/D: 350 deg (C=0.5, CHCl3)


Product Categories: Synthetic Organic Chemistry, Asymmetric Synthesis, Ligands
Other Product Names / Synonyms: (S)-(R)-BPPFA

Formula: C₃₈H₃₇FeNP₂
MW: 625.51 g/mol
Melting Pt: 142 °C
MDL Number: MFCD00075283
CAS Number: 55650-59-4
https://us.vwr.com/stibo/web/std.lang.all/95/10/28979510.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR