3,5-Difluoroanisole ≥97.0% (by GC)

Supplier: TCI America

Synonyms: 1,3-Difluoro-5-methoxybenzene

D2463-10G
TCD2463-10GEA 115 USD
TCD2463-10G
3,5-Difluoroanisole ≥97.0% (by GC)
3,5-Difluoroanisole

CAS Number: 93343-10-3
MDL Number: MFCD00042560
Molecular Formula: C7H6F2O
Molecular Weight: 144.12
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 43
Specific Gravity (20/20): 1.23

Formula: C₇H₆F₂O
MW: 144.12 g/mol
Boiling Pt: 148 °C
Density: 1.23 (20 °C)
Flash Pt: 43 °C
MDL Number: MFCD00042560
CAS Number: 93343-10-3
UN: 1993
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/93/85/28979385.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR