Diphenyliodonium iodide ≥90.0% (by HPLC)

D2373-5G
TCD2373-5GEA 176.46 USD
TCD2373-5G
Diphenyliodonium iodide ≥90.0% (by HPLC)
Diphenyliodonium iodide

CAS Number: 2217-79-0
MDL Number: MFCD00011910
Molecular Formula: C12H10I2
Molecular Weight: 408.02
Purity/Analysis Method: 90.0% (HPLC)
Form: Crystal
Melting point (°C): 163

Formula: C₁₂H₁₀I₂
Melting Pt: 163 °C
MDL Number: MFCD00011910
CAS Number: 2217-79-0
UN: 2811
ADR: 6.1,III
https://us.vwr.com/stibo/web/std.lang.all/93/00/28979300.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR