Diethoxymethylvinylsilane ≥97.0% (by GC)

Supplier: TCI America

Synonyms: Diethoxy(methyl)(vinyl)silane

D2318-25ML
TCD2318-25MLEA 69.08 USD
TCD2318-25ML
Diethoxymethylvinylsilane ≥97.0% (by GC)
Diethoxymethylvinylsilane

CAS Number: 5507-44-8
MDL Number: MFCD00009067
Molecular Formula: C7H16O2Si
Molecular Weight: 160.29
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 134
Flash Point (°C): 17
Specific Gravity (20/20): 0.86


Product Categories: Materials Chemistry, Silane Coupling Agents
Other Product Names / Synonyms: Ethenyldiethoxymethylsilane

Formula: C₇H₁₆O₂Si
MW: 160.29 g/mol
Boiling Pt: 134 °C
Density: 0.86 (20 °C)
Flash Pt: 17 °C
MDL Number: MFCD00009067
CAS Number: 5507-44-8
https://us.vwr.com/stibo/web/std.lang.all/92/51/28979251.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR