3,3'-Difluorobiphenyl ≥98.0% (by GC)

Synonyms: 3,3'-Difluoro-1,1'-biphenyl

D2180-1G
TCD2180-1GEA 235.87 USD
TCD2180-1G
3,3'-Difluorobiphenyl ≥98.0% (by GC)
3,3'-Difluorobiphenyl

CAS Number: 396-64-5
MDL Number: MFCD00039216
Molecular Formula: C12H8F2
Molecular Weight: 190.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Freezing point (°C): 6
Specific Gravity (20/20): 1.21

Formula: C₁₂H₈F₂
MW: 190.19 g/mol
Boiling Pt: 130 °C
Density: 1,21 (20 °C)
MDL Number: MFCD00039216
CAS Number: 396-64-5

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR