2,5-Difluorophenol ≥95.0% (by GC)
Supplier: TCI America
D2036-5G
TCD2036-5GEA
135.89
USD
TCD2036-5G
2,5-Difluorophenol ≥95.0% (by GC)
2,5-Difluorophenol
CAS Number: 2713-31-7
MDL Number: MFCD00042501
Molecular Formula: C6H4F2O
Molecular Weight: 130.09
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 145
Melting point (°C): 42
Flash Point (°C): 53
Formula:
C₆H₄F₂O MW: 130.09 g/mol Boiling Pt: 145 °C Melting Pt: 42 °C Flash Pt: 53 °C |
MDL Number:
MFCD00042501 CAS Number: 2713-31-7 UN: 2921 ADR: 8,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...