3,5-Dichlorobenzaldehyde ≥97.0% (by GC)
Supplier: TCI America
D2012-5G
D2012-1G
TCD2012-1GEA
18.27
USD
TCD2012-1G
TCD2012-5G
3,5-Dichlorobenzaldehyde ≥97.0% (by GC)
3,5-Dichlorobenzaldehyde
CAS Number: 10203-08-4
MDL Number: MFCD00003352
Molecular Formula: C7H4Cl2O
Molecular Weight: 175.01
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 215
Melting point (°C): 65
Formula:
C₇H₄Cl₂O MW: 175.01 g/mol Boiling Pt: 215 °C Melting Pt: 65 °C |
MDL Number:
MFCD00003352 CAS Number: 10203-08-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...