Dichlorodiethylsilane ≥98.0% (by GC)

Supplier: TCI America

D1976-25G D1976-5G
TCD1976-5GEA 57 USD
TCD1976-5G TCD1976-25G
Dichlorodiethylsilane ≥98.0% (by GC)
Dichlorodiethylsilane

CAS Number: 1719-53-5
MDL Number: MFCD00000493
Molecular Formula: C4H10Cl2Si
Molecular Weight: 157.11
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 129
Melting point (°C): -97
Flash Point (°C): 24
Specific Gravity (20/20): 1.06

Formula: C₄H₁₀Cl₂Si
Boiling Pt: 129 °C
Melting Pt: –97 °C
Density: 1.06 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00000493
CAS Number: 1719-53-5
UN: 1767
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/87/72/28978772.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR