1,2-Dibromo-1-phenylethane ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (1,2-Dibromoethyl)benzene

D1914-25G
TCD1914-25GEA 232.16 USD
TCD1914-25G
1,2-Dibromo-1-phenylethane ≥98.0% (by GC)
1,2-Dibromo-1-phenylethane

CAS Number: 93-52-7
MDL Number: MFCD00000138
Molecular Formula: C8H8Br2
Molecular Weight: 263.96
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 89
Melting point (°C): 74


Other Product Names / Synonyms: Styrene Dibromide

Formula: C₆H₅CHBrCH₂Br
MW: 263.96 g/mol
Boiling Pt: 89 °C
Melting Pt: 74 °C
MDL Number: MFCD00000138
CAS Number: 93-52-7
UN: 3261
ADR: 8,III
https://us.vwr.com/stibo/web/std.lang.all/87/12/28978712.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR