1-Chloro-3,4-dinitrobenzene ≥90.0% (by GC)
Supplier: TCI America
D1900-25G
TCD1900-25GEA
105.56
USD
TCD1900-25G
1-Chloro-3,4-dinitrobenzene ≥90.0% (by GC)
1-Chloro-3,4-dinitrobenzene
CAS Number: 610-40-2
MDL Number: MFCD00007212
Molecular Formula: C6H3ClN2O4
Molecular Weight: 202.55
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 315
Melting point (°C): 36
Flash Point (°C): 110
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-Chloro-1,2-dinitrobenzene
Formula:
C₆H₃ClN₂O₄ MW: 202.55 g/mol Boiling Pt: 315 °C Melting Pt: 36 °C Flash Pt: 110 °C |
MDL Number:
MFCD00007212 CAS Number: 610-40-2 UN: 1577 ADR: 6.1,II |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...