Dibenzosuberenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 5H-Dibenzo[a,d]cyclohepten-5-one5H-Dibenzo[a,d][7]annulen-5-one

D1791-25G
TCD1791-25GEA 77.77 USD
TCD1791-25G
Dibenzosuberenone ≥98.0% (by GC)
Dibenzosuberenone

CAS Number: 2222-33-5
MDL Number: MFCD00003588
Molecular Formula: C15H10O
Molecular Weight: 206.24
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 210
Melting point (°C): 89


Other Product Names / Synonyms: 5H-Dibenzo[a,d]cyclohepten-5-one

Formula: C₁₅H₁₀O
MW: 206.24 g/mol
Boiling Pt: 210 °C
Melting Pt: 89 °C
MDL Number: MFCD00003588
CAS Number: 2222-33-5
https://us.vwr.com/stibo/web/std.lang.all/85/70/28978570.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR