(2,4-Dinitrophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D1768-25G
TCD1768-25GEA
60.9
USD
TCD1768-25G
(2,4-Dinitrophenyl)acetic acid ≥98.0% (by HPLC, titration analysis)
(2,4-Dinitrophenyl)acetic acid
CAS Number: 643-43-6
MDL Number: MFCD00007227
Molecular Formula: C8H6N2O6
Molecular Weight: 226.14
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 189
Formula:
(C₆H₃(NO₂)₂)CH₂COOH MW: 226.15 g/mol Melting Pt: 189 °C |
MDL Number:
MFCD00007227 CAS Number: 643-43-6 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...