Dicyclopropyl ketone ≥95.0% (by GC)
Supplier: TCI America
Synonyms:
Dicyclopropylketone
, Dicyclopropylmethanone
D1758-25ML
TCD1758-25MLEA
41.31
USD
TCD1758-25ML
Dicyclopropyl ketone ≥95.0% (by GC)
Dicyclopropyl ketone
CAS Number: 1121-37-5
MDL Number: MFCD00001276
Molecular Formula: C7H10O
Molecular Weight: 110.16
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 67
Flash Point (°C): 39
Specific Gravity (20/20): 0.97
Formula:
C₇H₁₀O MW: 110.16 g/mol Boiling Pt: 67 °C Density: 0.97 (20 °C) Flash Pt: 39 °C |
MDL Number:
MFCD00001276 CAS Number: 1121-37-5 UN: 1224 ADR: 3,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...