α-Bromo-2,6-dichlorotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: alpha-Bromo-2,6-dichlorotoluene2,6-Dichlorobenzyl bromide

D1749-25G
TCD1749-25GEA 54.81 USD
TCD1749-25G
α-Bromo-2,6-dichlorotoluene ≥98.0% (by GC)
α-Bromo-2,6-dichlorotoluene

CAS Number: 20443-98-5
MDL Number: MFCD00000577
Molecular Formula: C7H5BrCl2
Molecular Weight: 239.92
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 57
Flash Point (°C): 112


Other Product Names / Synonyms: alpha-Bromo-2,6-dichlorotoluene

Formula: C₇H₅BrCl₂
MW: 239.93 g/mol
Boiling Pt: 135 °C
Melting Pt: 57 °C
Flash Pt: 112 °C
MDL Number: MFCD00000577
CAS Number: 20443-98-5
UN: 3261
ADR: 8,II
https://us.vwr.com/stibo/web/std.lang.all/85/30/28978530.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR