Decahydronaphthalene (mixture of cis and trans isomers) ≥99.0% (by GC)

Supplier: TCI America

Synonyms: DecalinBicyclo[4.4.0]decaneDecahydronaphthalene

D1738-500ML
TCD1738-500MLEA 45.78 USD
TCD1738-500ML
Decahydronaphthalene (mixture of cis and trans isomers) ≥99.0% (by GC)
Decahydronaphthalene (mixture of cis and trans isomers)

[Testing Methods for Sulfur in Crude Oil and Petroleum Products]
CAS Number: 91-17-8
MDL Number: MFCD00004130
Molecular Formula: C10H18
Molecular Weight: 138.25
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 190
Flash Point (°C): 58
Specific Gravity (20/20): 0.88

Formula: C₁₀H₁₈
MW: 138.25 g/mol
Boiling Pt: 190 °C
Density: 0.88 (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00004130
CAS Number: 91-17-8
UN: 1147
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/85/20/28978520.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR