(Dimethylamino)acetaldehyde diethylacetal ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,2-Diethoxy-N,N-dimethylethanamine

D1703-25ML
TCD1703-25MLEA 47.25 USD
TCD1703-25ML
(Dimethylamino)acetaldehyde diethylacetal ≥97.0% (by GC, titration analysis)
(Dimethylamino)acetaldehyde diethylacetal

CAS Number: 3616-56-6
MDL Number: MFCD00009232
Molecular Formula: C8H19NO2
Molecular Weight: 161.25
Purity/Analysis Method: 97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 45
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: N-(2,2-Diethoxyethyl)dimethylamine

Formula: C₈H₁₉NO₂
MW: 161.25 g/mol
Boiling Pt: 60 °C
Density: 0.87 (20 °C)
Flash Pt: 45 °C
MDL Number: MFCD00009232
CAS Number: 3616-56-6
https://us.vwr.com/stibo/web/std.lang.all/84/91/28978491.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR