4,4'-Dibromooctafluorobiphenyl (DBOB) ≥95.0% (by GC)

Supplier: TCI America

Synonyms: 4,4-Dibromooctafluorobiphenyl

D1696-1G
TCD1696-1GEA 42 USD
TCD1696-1G
4,4'-Dibromooctafluorobiphenyl (DBOB) ≥95.0% (by GC)
4,4'-Dibromooctafluorobiphenyl (DBOB)

CAS Number: 10386-84-2
MDL Number: MFCD00000310
Molecular Formula: C12Br2F8
Molecular Weight: 455.93
Purity/Analysis Method: 95.0% (GC)
Form: Crystal
Melting point (°C): 115

Formula: C₁₂Br₂F₈
MW: 455.94 g/mol
Melting Pt: 115 °C
MDL Number: MFCD00000310
CAS Number: 10386-84-2
https://us.vwr.com/stibo/web/std.lang.all/84/83/28978483.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR