2,5-Difluoroaniline ≥98.0% (by GC)
Supplier: TCI America
D1634-25G
TCD1634-25GEA
102.52
USD
TCD1634-25G
2,5-Difluoroaniline ≥98.0% (by GC)
2,5-Difluoroaniline
CAS Number: 367-30-6
MDL Number: MFCD00007651
Molecular Formula: C6H5F2N
Molecular Weight: 129.11
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 174
Flash Point (°C): 68
Freezing point (°C): 13
Specific Gravity (20/20): 1.29
Formula:
C₆H₅F₂N MW: 129.11 g/mol Boiling Pt: 174 °C Density: 1.29 (20 °C) Flash Pt: 68 °C |
MDL Number:
MFCD00007651 CAS Number: 367-30-6 UN: 2941 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...