α,2-Dibromotoluene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Bromobenzyl bromide

D1514-5G D1514-25G
TCD1514-25GEA 86.9 USD
TCD1514-25G TCD1514-5G
α,2-Dibromotoluene ≥98.0% (by GC)
α,2-Dibromotoluene

CAS Number: 3433-80-5
MDL Number: MFCD00000173
Molecular Formula: C7H6Br2
Molecular Weight: 249.93
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 30


Other Product Names / Synonyms: alpha,2-Dibromotoluene

Formula: C₇H₆Br₂
MW: 249.93 g/mol
Flash Pt: 110 °C
MDL Number: MFCD00000173
CAS Number: 3433-80-5
UN: 2293
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/83/29/28978329.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR